Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAAAAGCCTCTCACCTGGTATT[A/G]TCCCCTTTTATAACTTGTTTTTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606189 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRIM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRIM1 - cysteine rich transmembrane BMP regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016441.2 | Intron | NP_057525.1 | ||||
XM_005264357.4 | Intron | XP_005264414.1 | ||||
XM_011532898.2 | Intron | XP_011531200.1 | ||||
XM_011532899.2 | Intron | XP_011531201.1 | ||||
XM_011532901.2 | Intron | XP_011531203.1 | ||||
XM_017004258.1 | Intron | XP_016859747.1 | ||||
XM_017004259.1 | Intron | XP_016859748.1 | ||||
XM_017004260.1 | Intron | XP_016859749.1 | ||||
XM_017004261.1 | Intron | XP_016859750.1 | ||||
XM_017004262.1 | Intron | XP_016859751.1 |
LOC100288911 - uncharacterized LOC100288911 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |