Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNCRIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYNCRIP - synaptotagmin binding cytoplasmic RNA interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159673.1 | 3582 | UTR 3 | NP_001153145.1 | |||
NM_001159674.1 | 3582 | UTR 3 | NP_001153146.1 | |||
NM_001159675.1 | 3582 | Intron | NP_001153147.1 | |||
NM_001159676.1 | 3582 | UTR 3 | NP_001153148.1 | |||
NM_001159677.1 | 3582 | UTR 3 | NP_001153149.1 | |||
NM_001253771.1 | 3582 | UTR 3 | NP_001240700.1 | |||
NM_006372.4 | 3582 | Intron | NP_006363.4 | |||
XM_005248635.4 | 3582 | UTR 3 | XP_005248692.1 | |||
XM_005248636.2 | 3582 | Intron | XP_005248693.1 | |||
XM_005248637.2 | 3582 | Intron | XP_005248694.1 | |||
XM_017010176.1 | 3582 | UTR 3 | XP_016865665.1 | |||
XM_017010177.1 | 3582 | UTR 3 | XP_016865666.1 | |||
XM_017010178.1 | 3582 | UTR 3 | XP_016865667.1 | |||
XM_017010179.1 | 3582 | UTR 3 | XP_016865668.1 | |||
XM_017010180.1 | 3582 | Intron | XP_016865669.1 |