Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609644 MIM: 186947 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FANCM - Fanconi anemia complementation group M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FKBP3 - FK506 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002013.3 | Intron | NP_002004.1 | ||||
XM_011536565.2 | Intron | XP_011534867.1 | ||||
XM_017021092.1 | Intron | XP_016876581.1 | ||||
XM_017021093.1 | Intron | XP_016876582.1 |
Set Membership: |
HapMap |