Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606775 MIM: 601732 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSPG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSPG5 - chondroitin sulfate proteoglycan 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCC1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003074.3 | Intron | NP_003065.3 | ||||
XM_011534034.1 | Intron | XP_011532336.1 |
Set Membership: |
HapMap |