Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC553103 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC553103 - uncharacterized LOC553103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3936 - microRNA 3936 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A5 - solute carrier family 22 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308122.1 | Intron | NP_001295051.1 | ||||
NM_003060.3 | Intron | NP_003051.1 | ||||
XM_011543590.2 | Intron | XP_011541892.1 | ||||
XM_017009778.1 | Intron | XP_016865267.1 |