Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615117 MIM: 601788 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf88 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C2orf88 - chromosome 2 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042519.1 | Intron | NP_001035984.1 | ||||
NM_001042520.1 | Intron | NP_001035985.1 | ||||
NM_001042521.1 | Intron | NP_001035986.1 | ||||
NM_032321.2 | Intron | NP_115697.2 | ||||
XM_005246905.1 | Intron | XP_005246962.1 | ||||
XM_006712795.2 | Intron | XP_006712858.1 | ||||
XM_011511982.1 | Intron | XP_011510284.1 | ||||
XM_011511983.1 | Intron | XP_011510285.1 | ||||
XM_011511984.2 | Intron | XP_011510286.1 | ||||
XM_011511985.2 | Intron | XP_011510287.1 | ||||
XM_011511986.2 | Intron | XP_011510288.1 | ||||
XM_017005096.1 | Intron | XP_016860585.1 |
MSTN - myostatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005259.2 | Intron | NP_005250.1 |
Set Membership: |
HapMap |