Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604190 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC553103 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC553103 - uncharacterized LOC553103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A4 - solute carrier family 22 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003059.2 | Intron | NP_003050.2 | ||||
XM_006714675.3 | Intron | XP_006714738.1 | ||||
XM_011543589.2 | Intron | XP_011541891.1 | ||||
XM_017009776.1 | Intron | XP_016865265.1 |
Set Membership: |
HapMap |