Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NALCN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NALCN - sodium leak channel, non-selective | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052867.2 | 6771 | UTR 3 | NP_443099.1 | |||
XM_011521067.2 | 6771 | UTR 3 | XP_011519369.1 | |||
XM_011521068.2 | 6771 | Intron | XP_011519370.1 | |||
XM_011521069.2 | 6771 | Intron | XP_011519371.1 | |||
XM_017020534.1 | 6771 | Intron | XP_016876023.1 | |||
XM_017020535.1 | 6771 | Intron | XP_016876024.1 | |||
XM_017020536.1 | 6771 | Intron | XP_016876025.1 | |||
XM_017020537.1 | 6771 | Intron | XP_016876026.1 |
NALCN-AS1 - NALCN antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |