Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613110 MIM: 603106 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLCAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLCAP - bladder cancer associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167820.1 | 1146 | UTR 3 | NP_001161292.1 | |||
NM_001167821.1 | 1146 | UTR 3 | NP_001161293.1 | |||
NM_001167822.2 | 1146 | UTR 3 | NP_001161294.1 | |||
NM_001167823.1 | 1146 | UTR 3 | NP_001161295.1 | |||
NM_001317074.1 | 1146 | UTR 3 | NP_001304003.1 | |||
NM_001317075.1 | 1146 | UTR 3 | NP_001304004.1 | |||
NM_006698.3 | 1146 | UTR 3 | NP_006689.1 | |||
XM_017027599.1 | 1146 | UTR 3 | XP_016883088.1 |
NNAT - neuronatin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |