Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612859 MIM: 606025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR568 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR568 - microRNA 568 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIGIT - T-cell immunoreceptor with Ig and ITIM domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173799.3 | 24857 | Intron | NP_776160.2 |
ZBTB20 - zinc finger and BTB domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164342.2 | 24857 | UTR 3 | NP_001157814.1 | |||
NM_001164343.2 | 24857 | UTR 3 | NP_001157815.1 | |||
NM_001164344.2 | 24857 | UTR 3 | NP_001157816.1 | |||
NM_001164345.2 | 24857 | UTR 3 | NP_001157817.1 | |||
NM_001164346.2 | 24857 | UTR 3 | NP_001157818.1 | |||
NM_001164347.2 | 24857 | UTR 3 | NP_001157819.1 | |||
NM_015642.5 | 24857 | UTR 3 | NP_056457.3 |