Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 610178 MIM: 607384 | ||||||||||||||||||||||||||
Literature Links: |
KIAA0586 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KIAA0586 - KIAA0586 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244189.1 | Intron | NP_001231118.1 | ||||
NM_001244190.1 | Intron | NP_001231119.1 | ||||
NM_001244191.1 | Intron | NP_001231120.1 | ||||
NM_001244192.1 | Intron | NP_001231121.1 | ||||
NM_001244193.1 | Intron | NP_001231122.1 |
TIMM9 - translocase of inner mitochondrial membrane 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |