Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606078 MIM: 610440 | ||||||||||||||||||||
Literature Links: |
MKL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MKL1 - megakaryoblastic leukemia (translocation) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282660.1 | 4298 | UTR 3 | NP_001269589.1 | |||
NM_001282661.1 | 4298 | UTR 3 | NP_001269590.1 | |||
NM_001282662.1 | 4298 | UTR 3 | NP_001269591.1 | |||
NM_001318139.1 | 4298 | UTR 3 | NP_001305068.1 | |||
NM_020831.4 | 4298 | UTR 3 | NP_065882.1 | |||
XM_005261692.1 | 4298 | UTR 3 | XP_005261749.1 | |||
XM_005261694.1 | 4298 | UTR 3 | XP_005261751.1 | |||
XM_011530283.1 | 4298 | UTR 3 | XP_011528585.1 | |||
XM_011530285.1 | 4298 | UTR 3 | XP_011528587.1 | |||
XM_011530287.1 | 4298 | UTR 3 | XP_011528589.1 | |||
XM_017028888.1 | 4298 | UTR 3 | XP_016884377.1 | |||
XM_017028889.1 | 4298 | UTR 3 | XP_016884378.1 |
SGSM3 - small G protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301849.1 | 4298 | Intron | NP_001288778.1 | |||
NM_015705.5 | 4298 | Intron | NP_056520.2 | |||
XM_005261572.2 | 4298 | Intron | XP_005261629.1 | |||
XM_005261573.3 | 4298 | Intron | XP_005261630.1 | |||
XM_005261574.4 | 4298 | Intron | XP_005261631.1 | |||
XM_005261575.1 | 4298 | Intron | XP_005261632.1 | |||
XM_011530158.1 | 4298 | Intron | XP_011528460.1 | |||
XM_011530159.1 | 4298 | Intron | XP_011528461.1 | |||
XM_011530160.1 | 4298 | Intron | XP_011528462.1 | |||
XM_011530161.1 | 4298 | Intron | XP_011528463.1 | |||
XM_011530162.2 | 4298 | Intron | XP_011528464.1 | |||
XM_017028775.1 | 4298 | Intron | XP_016884264.1 | |||
XM_017028776.1 | 4298 | Intron | XP_016884265.1 | |||
XM_017028777.1 | 4298 | Intron | XP_016884266.1 | |||
XM_017028778.1 | 4298 | Intron | XP_016884267.1 | |||
XM_017028779.1 | 4298 | Intron | XP_016884268.1 | |||
XM_017028780.1 | 4298 | Intron | XP_016884269.1 |