Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607430 MIM: 601240 | ||||||||||||||||||||
Literature Links: |
DAZAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAZAP1 - DAZ associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018959.3 | Intron | NP_061832.2 | ||||
NM_170711.2 | Intron | NP_733829.1 | ||||
XM_005259531.3 | Intron | XP_005259588.1 | ||||
XM_005259534.3 | Intron | XP_005259591.1 | ||||
XM_005259535.3 | Intron | XP_005259592.1 | ||||
XM_005259536.3 | Intron | XP_005259593.1 | ||||
XM_011527904.2 | Intron | XP_011526206.1 | ||||
XM_011527905.2 | Intron | XP_011526207.1 | ||||
XM_011527906.2 | Intron | XP_011526208.1 | ||||
XM_011527907.2 | Intron | XP_011526209.1 | ||||
XM_011527908.2 | Intron | XP_011526210.1 | ||||
XM_011527909.2 | Intron | XP_011526211.1 | ||||
XM_011527910.2 | Intron | XP_011526212.1 | ||||
XM_017026581.1 | Intron | XP_016882070.1 | ||||
XM_017026582.1 | Intron | XP_016882071.1 |
GAMT - guanidinoacetate N-methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |