Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607679 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOCK4 - dedicator of cytokinesis 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014705.3 | 7409 | UTR 3 | NP_055520.3 | |||
XM_006716188.2 | 7409 | UTR 3 | XP_006716251.1 | |||
XM_006716189.2 | 7409 | UTR 3 | XP_006716252.1 | |||
XM_017012819.1 | 7409 | UTR 3 | XP_016868308.1 | |||
XM_017012820.1 | 7409 | UTR 3 | XP_016868309.1 | |||
XM_017012821.1 | 7409 | UTR 3 | XP_016868310.1 | |||
XM_017012822.1 | 7409 | UTR 3 | XP_016868311.1 | |||
XM_017012823.1 | 7409 | UTR 3 | XP_016868312.1 | |||
XM_017012824.1 | 7409 | UTR 3 | XP_016868313.1 | |||
XM_017012825.1 | 7409 | UTR 3 | XP_016868314.1 |
Set Membership: |
HapMap |