Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603158 | ||||||||||||||||||||
Literature Links: |
USP50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP50 - ubiquitin specific peptidase 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203494.4 | 5055 | Intron | NP_987090.2 |
USP8 - ubiquitin specific peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128610.2 | 5055 | UTR 3 | NP_001122082.1 | |||
NM_001283049.1 | 5055 | UTR 3 | NP_001269978.1 | |||
NM_005154.4 | 5055 | UTR 3 | NP_005145.3 | |||
XM_006720761.3 | 5055 | UTR 3 | XP_006720824.1 | |||
XM_006720762.3 | 5055 | Intron | XP_006720825.1 | |||
XM_011522193.2 | 5055 | Intron | XP_011520495.1 | |||
XM_017022718.1 | 5055 | Intron | XP_016878207.1 | |||
XM_017022719.1 | 5055 | Intron | XP_016878208.1 | |||
XM_017022720.1 | 5055 | Intron | XP_016878209.1 | |||
XM_017022721.1 | 5055 | Intron | XP_016878210.1 | |||
XM_017022722.1 | 5055 | Intron | XP_016878211.1 |