Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX4 - DEAD-box helicase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142549.1 | Intron | NP_001136021.1 | ||||
NM_001166533.1 | Intron | NP_001160005.1 | ||||
NM_001166534.1 | Intron | NP_001160006.1 | ||||
NM_024415.2 | Intron | NP_077726.1 | ||||
XM_011543495.2 | Intron | XP_011541797.1 | ||||
XM_011543497.2 | Intron | XP_011541799.1 |