Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600608 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P4HA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
P4HA2 - prolyl 4-hydroxylase subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017973.1 | 2067 | Intron | NP_001017973.1 | |||
NM_001017974.1 | 2067 | Intron | NP_001017974.1 | |||
NM_001142598.1 | 2067 | Intron | NP_001136070.1 | |||
NM_001142599.1 | 2067 | Intron | NP_001136071.1 | |||
NM_004199.2 | 2067 | Intron | NP_004190.1 | |||
XM_005272116.4 | 2067 | UTR 3 | XP_005272173.1 | |||
XM_005272117.4 | 2067 | UTR 3 | XP_005272174.1 | |||
XM_005272118.4 | 2067 | UTR 3 | XP_005272175.1 | |||
XM_005272119.4 | 2067 | UTR 3 | XP_005272176.1 | |||
XM_005272120.4 | 2067 | UTR 3 | XP_005272177.1 | |||
XM_006714728.3 | 2067 | UTR 3 | XP_006714791.1 | |||
XM_006714729.3 | 2067 | UTR 3 | XP_006714792.1 | |||
XM_006714730.3 | 2067 | UTR 3 | XP_006714793.1 | |||
XM_011543705.2 | 2067 | UTR 3 | XP_011542007.1 | |||
XM_017010011.1 | 2067 | UTR 3 | XP_016865500.1 | |||
XM_017010012.1 | 2067 | UTR 3 | XP_016865501.1 |
P4HA2-AS1 - P4HA2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |