Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAGATCCACACCAATCACTGCC[T/C]GGAATGTGTGCGCGTGTGGCCGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611328 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJB5 - DnaJ heat shock protein family (Hsp40) member B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135004.2 | 180 | Intron | NP_001128476.2 | |||
NM_001135005.2 | 180 | Intron | NP_001128477.1 | |||
NM_012266.5 | 180 | Intron | NP_036398.3 | |||
XM_005251428.3 | 180 | Intron | XP_005251485.1 | |||
XM_006716751.2 | 180 | Intron | XP_006716814.1 | |||
XM_011517842.2 | 180 | UTR 5 | XP_011516144.1 |
DNAJB5-AS1 - DNAJB5 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF24 - PHD finger protein 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |