Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611898 MIM: 616999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENGASE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENGASE - endo-beta-N-acetylglucosaminidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBFOX3 - RNA binding protein, fox-1 homolog 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082575.2 | Intron | NP_001076044.1 | ||||
XM_011524359.1 | Intron | XP_011522661.1 | ||||
XM_011524360.1 | Intron | XP_011522662.1 | ||||
XM_011524362.2 | Intron | XP_011522664.1 | ||||
XM_011524363.2 | Intron | XP_011522665.1 | ||||
XM_011524365.2 | Intron | XP_011522667.1 | ||||
XM_011524366.2 | Intron | XP_011522668.1 | ||||
XM_011524367.2 | Intron | XP_011522669.1 | ||||
XM_017024208.1 | Intron | XP_016879697.1 | ||||
XM_017024209.1 | Intron | XP_016879698.1 | ||||
XM_017024210.1 | Intron | XP_016879699.1 | ||||
XM_017024211.1 | Intron | XP_016879700.1 |