Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138570 MIM: 604788 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GYS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GYS1 - glycogen synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161587.1 | 82 | Intron | NP_001155059.1 | |||
NM_002103.4 | 82 | Intron | NP_002094.2 |
RUVBL2 - RuvB like AAA ATPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321190.1 | 82 | UTR 5 | NP_001308119.1 | |||
NM_001321191.1 | 82 | UTR 5 | NP_001308120.1 | |||
NM_006666.2 | 82 | UTR 5 | NP_006657.1 | |||
XM_011526330.1 | 82 | Intron | XP_011524632.1 |