Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604879 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC12A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC12A7 - solute carrier family 12 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006598.2 | 3875 | UTR 3 | NP_006589.2 | |||
XM_005248231.3 | 3875 | UTR 3 | XP_005248288.1 | |||
XM_011513939.2 | 3875 | UTR 3 | XP_011512241.1 | |||
XM_011513940.2 | 3875 | UTR 3 | XP_011512242.1 | |||
XM_011513941.2 | 3875 | UTR 3 | XP_011512243.2 | |||
XM_017008958.1 | 3875 | UTR 3 | XP_016864447.1 | |||
XM_017008959.1 | 3875 | UTR 3 | XP_016864448.1 |
Set Membership: |
HapMap |