Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138248 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIA1 - glutamate ionotropic receptor AMPA type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000827.3 | 73 | Intron | NP_000818.2 | |||
NM_001114183.1 | 73 | Intron | NP_001107655.1 | |||
NM_001258019.1 | 73 | Intron | NP_001244948.1 | |||
NM_001258020.1 | 73 | Intron | NP_001244949.1 | |||
NM_001258021.1 | 73 | Missense Mutation | AGC,GGC | S,G 16 | NP_001244950.1 | |
NM_001258022.1 | 73 | Missense Mutation | AGC,GGC | S,G 16 | NP_001244951.1 | |
NM_001258023.1 | 73 | Intron | NP_001244952.1 | |||
XM_011537635.2 | 73 | Intron | XP_011535937.1 | |||
XM_017009392.1 | 73 | Missense Mutation | AGC,GGC | S,G 16 | XP_016864881.1 | |
XM_017009393.1 | 73 | Intron | XP_016864882.1 |