Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612299 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COMMD9 - COMM domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101653.1 | 2729 | UTR 3 | NP_001095123.1 | |||
NM_001307932.1 | 2729 | UTR 3 | NP_001294861.1 | |||
NM_001307937.1 | 2729 | UTR 3 | NP_001294866.1 | |||
NM_014186.3 | 2729 | UTR 3 | NP_054905.2 | |||
XM_017017625.1 | 2729 | Intron | XP_016873114.1 |