Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606676 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPV2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRPV2 - transient receptor potential cation channel subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016113.4 | 77 | UTR 5 | NP_057197.2 | |||
XM_005256676.1 | 77 | UTR 5 | XP_005256733.1 | |||
XM_005256677.1 | 77 | UTR 5 | XP_005256734.1 | |||
XM_005256678.4 | 77 | UTR 5 | XP_005256735.1 | |||
XM_006721541.3 | 77 | UTR 5 | XP_006721604.1 | |||
XM_006721543.3 | 77 | UTR 5 | XP_006721606.1 | |||
XM_011523922.1 | 77 | UTR 5 | XP_011522224.1 | |||
XM_011523923.1 | 77 | UTR 5 | XP_011522225.1 | |||
XM_011523925.2 | 77 | Intron | XP_011522227.1 | |||
XM_017024730.1 | 77 | UTR 5 | XP_016880219.1 | |||
XM_017024731.1 | 77 | UTR 5 | XP_016880220.1 | |||
XM_017024732.1 | 77 | UTR 5 | XP_016880221.1 |
Set Membership: |
HapMap |