Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603071 MIM: 607908 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR17 - G protein-coupled receptor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161415.1 | Intron | NP_001154887.1 | ||||
NM_001161416.1 | Intron | NP_001154888.1 | ||||
NM_001161417.1 | Intron | NP_001154889.1 | ||||
NM_005291.2 | Intron | NP_005282.1 | ||||
XM_017003833.1 | Intron | XP_016859322.1 |
LIMS2 - LIM zinc finger domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136037.2 | Intron | NP_001129509.2 | ||||
NM_001161403.1 | Intron | NP_001154875.1 | ||||
NM_001161404.1 | Intron | NP_001154876.1 | ||||
NM_001256542.1 | Intron | NP_001243471.1 | ||||
NM_017980.4 | Intron | NP_060450.2 | ||||
XM_005263710.2 | Intron | XP_005263767.1 | ||||
XM_006712627.3 | Intron | XP_006712690.1 | ||||
XM_006712628.3 | Intron | XP_006712691.1 | ||||
XM_011511453.1 | Intron | XP_011509755.1 | ||||
XM_017004469.1 | Intron | XP_016859958.1 |