Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOTCH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOTCH2 - notch 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200001.1 | 10302 | Intron | NP_001186930.1 | |||
NM_024408.3 | 10302 | UTR 3 | NP_077719.2 | |||
XM_005270901.3 | 10302 | UTR 3 | XP_005270958.1 | |||
XM_011541519.2 | 10302 | UTR 3 | XP_011539821.1 | |||
XM_011541520.2 | 10302 | UTR 3 | XP_011539822.1 | |||
XM_017001372.1 | 10302 | UTR 3 | XP_016856861.1 | |||
XM_017001373.1 | 10302 | UTR 3 | XP_016856862.1 |