Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608948 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZIC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZIC4 - Zic family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168378.1 | 2628 | UTR 3 | NP_001161850.1 | |||
NM_001168379.1 | 2628 | UTR 3 | NP_001161851.1 | |||
NM_001243256.1 | 2628 | UTR 3 | NP_001230185.1 | |||
NM_032153.5 | 2628 | UTR 3 | NP_115529.2 | |||
XM_017007314.1 | 2628 | Intron | XP_016862803.1 |
Set Membership: |
HapMap |