Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614758 MIM: 612430 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTN4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCTN4 - dynactin subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135643.1 | 1889 | UTR 3 | NP_001129115.1 | |||
NM_001135644.1 | 1889 | UTR 3 | NP_001129116.1 | |||
NM_016221.3 | 1889 | UTR 3 | NP_057305.1 | |||
XM_005268457.1 | 1889 | UTR 3 | XP_005268514.1 | |||
XM_011537644.1 | 1889 | UTR 3 | XP_011535946.1 | |||
XM_011537645.1 | 1889 | UTR 3 | XP_011535947.1 | |||
XM_011537646.1 | 1889 | UTR 3 | XP_011535948.1 | |||
XM_011537647.1 | 1889 | UTR 3 | XP_011535949.1 |
RBM22 - RNA binding motif protein 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |