Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611814 MIM: 616799 | ||||||||||||||||||||
Literature Links: |
ELOVL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELOVL2 - ELOVL fatty acid elongase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017770.3 | 1583 | UTR 3 | NP_060240.3 | |||
XM_011514716.2 | 1583 | UTR 3 | XP_011513018.1 | |||
XM_011514717.2 | 1583 | UTR 3 | XP_011513019.1 | |||
XM_017010985.1 | 1583 | Intron | XP_016866474.1 |
SYCP2L - synaptonemal complex protein 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |