Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615570 MIM: 611129 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SFXN2 - sideroflexin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBP1L - WW domain binding protein 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083913.1 | 1120 | Intron | NP_001077382.1 | |||
NM_017787.4 | 1120 | Intron | NP_060257.4 | |||
XM_011539913.2 | 1120 | Intron | XP_011538215.1 | |||
XM_017016360.1 | 1120 | UTR 5 | XP_016871849.1 |