Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613868 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC14A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC14A1 - solute carrier family 14 member 1 (Kidd blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128588.3 | 95 | UTR 5 | NP_001122060.3 | |||
NM_001146036.2 | 95 | UTR 5 | NP_001139508.2 | |||
NM_001146037.1 | 95 | Intron | NP_001139509.1 | |||
NM_001308278.1 | 95 | UTR 5 | NP_001295207.1 | |||
NM_001308279.1 | 95 | UTR 5 | NP_001295208.1 | |||
NM_015865.6 | 95 | UTR 5 | NP_056949.4 | |||
XM_005258329.1 | 95 | UTR 5 | XP_005258386.1 | |||
XM_005258333.1 | 95 | UTR 5 | XP_005258390.1 | |||
XM_006722526.3 | 95 | UTR 5 | XP_006722589.1 | |||
XM_011526142.1 | 95 | Intron | XP_011524444.1 | |||
XM_011526144.1 | 95 | UTR 5 | XP_011524446.1 |
Set Membership: |
HapMap |