Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAATGAAGCAATAAAAGTTCTC[C/T]TTTGTACTTGATCTTGTTTGAATTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601679 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2I PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTF2I - general transcription factor IIi | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163636.2 | Intron | NP_001157108.1 | ||||
NM_001280800.1 | Intron | NP_001267729.1 | ||||
NM_001518.4 | Intron | NP_001509.3 | ||||
NM_032999.3 | Intron | NP_127492.1 | ||||
NM_033000.3 | Intron | NP_127493.1 | ||||
NM_033001.3 | Intron | NP_127494.1 |
LOC101926943 - uncharacterized LOC101926943 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |