Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602726 MIM: 607093 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCN6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLCN6 - chloride voltage-gated channel 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256959.1 | Intron | NP_001243888.1 | ||||
NM_001286.3 | Intron | NP_001277.1 |
MTHFR - methylenetetrahydrofolate reductase (NAD(P)H) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005957.4 | Intron | NP_005948.3 | ||||
XM_005263458.3 | Intron | XP_005263515.1 | ||||
XM_005263460.4 | Intron | XP_005263517.1 | ||||
XM_005263462.4 | Intron | XP_005263519.1 | ||||
XM_005263463.3 | Intron | XP_005263520.1 | ||||
XM_011541495.2 | Intron | XP_011539797.1 | ||||
XM_011541496.2 | Intron | XP_011539798.1 | ||||
XM_017001328.1 | Intron | XP_016856817.1 |
Set Membership: |
HapMap JSNP |