Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602878 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC30A3 - solute carrier family 30 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318949.1 | 1914 | UTR 3 | NP_001305878.1 | |||
NM_001318950.1 | 1914 | UTR 3 | NP_001305879.1 | |||
NM_001318951.1 | 1914 | UTR 3 | NP_001305880.1 | |||
NM_003459.4 | 1914 | UTR 3 | NP_003450.2 | |||
XM_005264547.1 | 1914 | UTR 3 | XP_005264604.1 | |||
XM_005264548.3 | 1914 | UTR 3 | XP_005264605.1 | |||
XM_006712100.2 | 1914 | UTR 3 | XP_006712163.1 | |||
XM_011533102.2 | 1914 | UTR 3 | XP_011531404.1 | |||
XM_011533103.2 | 1914 | UTR 3 | XP_011531405.1 | |||
XM_017004873.1 | 1914 | UTR 3 | XP_016860362.1 | |||
XM_017004874.1 | 1914 | UTR 3 | XP_016860363.1 | |||
XM_017004875.1 | 1914 | UTR 3 | XP_016860364.1 |
Set Membership: |
HapMap |