Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPP7 - membrane palmitoylated protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318170.1 | 3102 | UTR 3 | NP_001305099.1 | |||
NM_173496.4 | 3102 | UTR 3 | NP_775767.2 | |||
XM_005252369.3 | 3102 | Intron | XP_005252426.1 | |||
XM_011519337.2 | 3102 | UTR 3 | XP_011517639.1 | |||
XM_011519338.2 | 3102 | UTR 3 | XP_011517640.1 | |||
XM_017015741.1 | 3102 | UTR 3 | XP_016871230.1 | |||
XM_017015742.1 | 3102 | UTR 3 | XP_016871231.1 | |||
XM_017015743.1 | 3102 | Intron | XP_016871232.1 |