Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606167 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR132 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GPR132 - G protein-coupled receptor 132 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278694.1 | 969 | Silent Mutation | AAC,AAT | N,N 277 | NP_001265623.1 | |
NM_001278695.1 | 969 | Silent Mutation | AAC,AAT | N,N 268 | NP_001265624.1 | |
NM_001278696.1 | 969 | Silent Mutation | AAC,AAT | N,N 89 | NP_001265625.1 | |
NM_013345.3 | 969 | Silent Mutation | AAC,AAT | N,N 277 | NP_037477.1 |
Set Membership: |
HapMap |