Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616701 MIM: 608844 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
COMMD4 - COMM domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284377.1 | 686 | UTR 3 | NP_001271306.1 | |||
NM_001284378.1 | 686 | UTR 3 | NP_001271307.1 | |||
NM_001284379.1 | 686 | UTR 3 | NP_001271308.1 | |||
NM_001321844.1 | 686 | UTR 3 | NP_001308773.1 | |||
NM_001321845.1 | 686 | UTR 3 | NP_001308774.1 | |||
NM_001321846.1 | 686 | UTR 3 | NP_001308775.1 | |||
NM_001321847.1 | 686 | UTR 3 | NP_001308776.1 | |||
NM_001321848.1 | 686 | UTR 3 | NP_001308777.1 | |||
NM_017828.4 | 686 | UTR 3 | NP_060298.2 | |||
XM_005254509.3 | 686 | UTR 3 | XP_005254566.1 | |||
XM_005254510.3 | 686 | UTR 3 | XP_005254567.1 | |||
XM_011521739.1 | 686 | UTR 3 | XP_011520041.1 | |||
XM_011521740.1 | 686 | UTR 3 | XP_011520042.1 | |||
XM_011521741.1 | 686 | UTR 3 | XP_011520043.1 | |||
XM_017022385.1 | 686 | UTR 3 | XP_016877874.1 |
NEIL1 - nei like DNA glycosylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |