Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613127 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRDL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHRDL2 - chordin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278473.2 | Intron | NP_001265402.1 | ||||
NM_001304390.1 | Intron | NP_001291319.1 | ||||
NM_001304391.1 | Intron | NP_001291320.1 | ||||
NM_001304415.1 | Intron | NP_001291344.1 | ||||
NM_001304416.1 | Intron | NP_001291345.1 | ||||
NM_001304417.1 | Intron | NP_001291346.1 | ||||
NM_015424.5 | Intron | NP_056239.3 |