Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604646 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0930 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA0930 - KIAA0930 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009880.1 | 4980 | UTR 3 | NP_001009880.1 | |||
NM_015264.1 | 4980 | UTR 3 | NP_056079.1 | |||
XM_011530017.2 | 4980 | UTR 3 | XP_011528319.1 | |||
XM_017028672.1 | 4980 | UTR 3 | XP_016884161.1 |
MIR1249 - microRNA 1249 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP50 - nucleoporin 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |