Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608780 MIM: 614725 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2H5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GTF2H5 - general transcription factor IIH subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207118.2 | 78 | UTR 5 | NP_997001.1 | |||
XM_017010862.1 | 78 | Silent Mutation | CCC,CCG | P,P 21 | XP_016866351.1 |
SERAC1 - serine active site containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032861.3 | 78 | Intron | NP_116250.3 | |||
XM_006715586.2 | 78 | Intron | XP_006715649.1 | |||
XM_011536196.2 | 78 | Intron | XP_011534498.1 | |||
XM_011536198.2 | 78 | Intron | XP_011534500.1 |
Set Membership: |
HapMap |