Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604821 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LILRB4 - leukocyte immunoglobulin like receptor B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278426.3 | 698 | Silent Mutation | TTC,TTT | F,F 153 | NP_001265355.2 | |
NM_001278427.3 | 698 | Silent Mutation | TTC,TTT | F,F 153 | NP_001265356.2 | |
NM_001278428.3 | 698 | Silent Mutation | TTC,TTT | F,F 153 | NP_001265357.2 | |
NM_001278429.3 | 698 | Silent Mutation | TTC,TTT | F,F 182 | NP_001265358.2 | |
NM_001278430.3 | 698 | Intron | NP_001265359.2 | |||
XM_017026215.1 | 698 | Silent Mutation | TTC,TTT | F,F 194 | XP_016881704.1 | |
XM_017026216.1 | 698 | Silent Mutation | TTC,TTT | F,F 194 | XP_016881705.1 | |
XM_017026217.1 | 698 | Silent Mutation | TTC,TTT | F,F 194 | XP_016881706.1 |
Set Membership: |
HapMap |