Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609534 MIM: 614853 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATAD5 - ATPase family, AAA domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024857.4 | 658 | Missense Mutation | CCT,TCT | P,S 87 | NP_079133.3 | |
XM_011525269.2 | 658 | Missense Mutation | CCT,TCT | P,S 87 | XP_011523571.1 | |
XM_011525270.2 | 658 | Missense Mutation | CCT,TCT | P,S 87 | XP_011523572.1 | |
XM_011525273.2 | 658 | Missense Mutation | CCT,TCT | P,S 87 | XP_011523575.1 | |
XM_011525274.2 | 658 | Intron | XP_011523576.1 | |||
XM_017025111.1 | 658 | Missense Mutation | CCT,TCT | P,S 73 | XP_016880600.1 |
CRLF3 - cytokine receptor like factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |