Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL2 - kelch like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161521.1 | Intron | NP_001154993.1 | ||||
NM_001161522.1 | Intron | NP_001154994.1 | ||||
NM_007246.3 | Intron | NP_009177.3 | ||||
XM_011531572.2 | Intron | XP_011529874.1 | ||||
XM_011531575.2 | Intron | XP_011529877.1 | ||||
XM_011531576.2 | Intron | XP_011529878.1 | ||||
XM_017007674.1 | Intron | XP_016863163.1 | ||||
XM_017007675.1 | Intron | XP_016863164.1 | ||||
XM_017007676.1 | Intron | XP_016863165.1 | ||||
XM_017007677.1 | Intron | XP_016863166.1 |