Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612402 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ALS2CL - ALS2 C-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190707.1 | 4132 | UTR 3 | NP_001177636.1 | |||
NM_147129.4 | 4132 | UTR 3 | NP_667340.2 | |||
XM_005265025.1 | 4132 | UTR 3 | XP_005265082.1 | |||
XM_006713091.2 | 4132 | Intron | XP_006713154.1 | |||
XM_006713093.3 | 4132 | Intron | XP_006713156.1 | |||
XM_006713094.3 | 4132 | Intron | XP_006713157.1 | |||
XM_011533572.2 | 4132 | Intron | XP_011531874.1 | |||
XM_017006120.1 | 4132 | Intron | XP_016861609.1 |