Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
80 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606509 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCRL2 - Fc receptor like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159488.1 | Intron | NP_001152960.1 | ||||
NM_030764.3 | Intron | NP_110391.2 | ||||
XM_006711535.3 | Intron | XP_006711598.1 | ||||
XM_011509974.2 | Intron | XP_011508276.1 | ||||
XM_011509975.2 | Intron | XP_011508277.1 | ||||
XM_011509976.2 | Intron | XP_011508278.1 | ||||
XM_017002316.1 | Intron | XP_016857805.1 | ||||
XM_017002317.1 | Intron | XP_016857806.1 | ||||
XM_017002318.1 | Intron | XP_016857807.1 | ||||
XM_017002319.1 | Intron | XP_016857808.1 |
Set Membership: |
HapMap JSNP |