Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614264 MIM: 609607 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP30 - Rho GTPase activating protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025598.1 | Intron | NP_001020769.1 | ||||
NM_001287600.1 | Intron | NP_001274529.1 | ||||
NM_001287602.1 | Intron | NP_001274531.1 | ||||
NM_181720.2 | Intron | NP_859071.2 | ||||
XM_005245070.2 | Intron | XP_005245127.1 | ||||
XM_005245073.3 | Intron | XP_005245130.1 | ||||
XM_011509391.2 | Intron | XP_011507693.1 | ||||
XM_017000960.1 | Intron | XP_016856449.1 |
NECTIN4 - nectin cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030916.2 | Intron | NP_112178.2 | ||||
XM_005245508.3 | Intron | XP_005245565.1 | ||||
XM_011510021.2 | Intron | XP_011508323.1 | ||||
XM_011510022.2 | Intron | XP_011508324.1 | ||||
XM_011510023.1 | Intron | XP_011508325.1 |
Set Membership: |
HapMap JSNP |