Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGGAGGCGGGTGGTGCTCATGC[A/G]TGCCAGTGAGGGAGGGGCCTCGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616475 MIM: 608773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP72 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP72 - centrosomal protein 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018140.3 | 4429 | Intron | NP_060610.2 | |||
XM_005248322.3 | 4429 | Intron | XP_005248379.1 | |||
XM_011514063.1 | 4429 | Intron | XP_011512365.1 | |||
XM_011514064.2 | 4429 | Intron | XP_011512366.1 | |||
XM_017009626.1 | 4429 | Intron | XP_016865115.1 | |||
XM_017009627.1 | 4429 | Intron | XP_016865116.1 |
LOC101929895 - uncharacterized LOC101929895 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPPP - tubulin polymerization promoting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007030.2 | 4429 | UTR 3 | NP_008961.1 | |||
XM_005248237.3 | 4429 | UTR 3 | XP_005248294.2 | |||
XM_017008993.1 | 4429 | UTR 3 | XP_016864482.1 | |||
XM_017008994.1 | 4429 | UTR 3 | XP_016864483.1 |