Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616236 MIM: 611865 | ||||||||||||||||||||
Literature Links: |
CHADL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHADL - chondroadherin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EP300-AS1 - EP300 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
L3MBTL2 - L3MBTL2 polycomb repressive complex 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031488.4 | 118 | UTR 5 | NP_113676.2 | |||
XM_011530420.2 | 118 | Silent Mutation | GGA,GGG | G,G 9 | XP_011528722.1 | |
XM_011530421.2 | 118 | UTR 5 | XP_011528723.1 | |||
XM_017028976.1 | 118 | UTR 5 | XP_016884465.1 | |||
XM_017028977.1 | 118 | Intron | XP_016884466.1 | |||
XM_017028978.1 | 118 | Intron | XP_016884467.1 | |||
XM_017028979.1 | 118 | UTR 5 | XP_016884468.1 | |||
XM_017028980.1 | 118 | UTR 5 | XP_016884469.1 |