Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610703 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSPH1 - heat shock protein family H (Hsp110) member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286503.1 | Intron | NP_001273432.1 | ||||
NM_001286504.1 | Intron | NP_001273433.1 | ||||
NM_001286505.1 | Intron | NP_001273434.1 | ||||
NM_006644.3 | Intron | NP_006635.2 | ||||
XM_005266236.1 | Intron | XP_005266293.1 | ||||
XM_011534887.2 | Intron | XP_011533189.1 | ||||
XM_011534888.1 | Intron | XP_011533190.1 | ||||
XM_017020361.1 | Intron | XP_016875850.1 | ||||
XM_017020362.1 | Intron | XP_016875851.1 | ||||
XM_017020363.1 | Intron | XP_016875852.1 | ||||
XM_017020364.1 | Intron | XP_016875853.1 |