Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616639 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRDM8 - PR domain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099403.1 | 132 | Intron | NP_001092873.1 | |||
NM_020226.3 | 132 | Intron | NP_064611.3 | |||
XM_005263144.2 | 132 | Intron | XP_005263201.1 | |||
XM_005263145.2 | 132 | Intron | XP_005263202.1 | |||
XM_005263146.4 | 132 | Intron | XP_005263203.1 | |||
XM_011532133.2 | 132 | UTR 5 | XP_011530435.1 | |||
XM_011532135.2 | 132 | UTR 5 | XP_011530437.1 | |||
XM_011532140.2 | 132 | Intron | XP_011530442.1 | |||
XM_011532141.2 | 132 | Intron | XP_011530443.1 | |||
XM_017008468.1 | 132 | Intron | XP_016863957.1 | |||
XM_017008469.1 | 132 | Intron | XP_016863958.1 | |||
XM_017008470.1 | 132 | Intron | XP_016863959.1 |
Set Membership: |
HapMap |